2-Amino-9H-pyrido[2,3-b]indole ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
9H-Pyrido[2,3-b]indol-2-amine, 9H-1,9-Diazafluoren-2-amine
A3209-25MG
A3209-5MG
TCIAA3209-25MGEA
4200
NOK
TCIAA3209-25MG
TCIAA3209-5MG
2-Amino-9H-pyrido[2,3-b]indole ≥98.0% (by GC)
2-Amino-9H-pyrido[2,3-b]indole
Formel:
C₁₁H₉N₃ MW: 183,21 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00210750 CAS-nummer: 26148-68-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 199.0 to 203.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...