4-(Aminomethyl)cyclohexanamine cis- and trans-mixture ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 1-Amino-4-(aminomethyl)cyclohexane

A3258-1G A3258-5G
TCIAA3258-1GEA 1400 NOK
TCIAA3258-1G TCIAA3258-5G
4-(Aminomethyl)cyclohexanamine cis- and trans-mixture ≥96.0% (by GC, titration analysis)
4-(Aminomethyl)cyclohexanamine
Formel: C₇H₁₆N₂
MW: 128,22 g/mol
Kokepunkt: 101°C
MDL nummer: MFCD00049306
CAS-nummer: 13338-82-4
FN: 2735
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 % (total of isomer)
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR