1-Benzhydrylazetidin-3-amine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Amino-1-(diphenylmethyl)azetidine
A3283-1G
A3283-5G
TCIAA3283-1GEA
540
NOK
TCIAA3283-1G
TCIAA3283-5G
1-Benzhydrylazetidin-3-amine ≥98.0% (by GC)
1-Benzhydrylazetidin-3-amine
Formel:
C₁₆H₁₈N₂ MW: 238,34 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD03093386 CAS-nummer: 40432-52-8 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 61.000 to 65.000 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...