3-[(Allyloxy)methyl]-3-ethyloxetane ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Ethyl-3-(prop-2-enoxymethyl)oxetane

A3362-1G A3362-5G
TCIAA3362-1GEA 1175 NOK
TCIAA3362-1G TCIAA3362-5G
3-[(Allyloxy)methyl]-3-ethyloxetane ≥98.0% (by GC)
3-[(Allyloxy)methyl]-3-ethyloxetane
Formel: C₉H₁₆O₂
MW: 156,23 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD17012910
CAS-nummer: 3207-04-3
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR