Anethole trithione ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Felviten

A3364-25G A3364-5G
TCIAA3364-25GEA 3125 NOK
TCIAA3364-25G TCIAA3364-5G
Anethole trithione ≥98.0% (by GC)
Anethole trithione
Formel: C₁₀H₈OS₃
MW: 240,3649 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00129751
CAS-nummer: 532-11-6
EINECS: 208-528-5
Merck Indeks: 14,00644

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR