Dimethylsulphonazo III for spektrofotometri

Leverandør: TCI

Synonymer: 2,7-Bis(2-sulpho-p-tolylazo)-1,8-dihydroxy-3,6-naphthalenedisulphonic acid, 2,7-Bis[(4-methyl-2-sulphophenyl)azo]-1,8-dihydroxynaphthalene-3,6-disulphonic acid

A5008-1G A5008-5G
TCIAA5008-1GEA 865 NOK
TCIAA5008-1G TCIAA5008-5G
Dimethylsulphonazo III for spektrofotometri
Dimethylsulphonazo III

Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba

Formel: C₂₄H₂₀N₄O₁₄S₄
MW: 716,68 g/mol
MDL nummer: MFCD00151118
CAS-nummer: 14979-11-4
EINECS: 239-057-3

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 570.0 to 580.0 nm (H2O)
Absorbance (E1%1cm) min. 400 (H2O, 570.0 to 580.0 nm)
Drying loss max. 15.0 %
Ignition residue (Sulfate) max. 7.0 % (after drying)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR