2,3-Naphthalenedialdehyde (NDA) ≥99.0% (by GC) fluorimetric reagent for primary amines
Leverandør: TCI
|
Warning
|
Synonymer:
Naphthalene-2,3-dicarbaldehyde, Naphthalene-2,3-dicarboxaldehyde, NDA
A5594-1G
A5594-100MG
TCIAA5594-1GEA
9825
NOK
TCIAA5594-1G
TCIAA5594-100MG
2,3-Naphthalenedialdehyde (NDA) ≥99.0% (by GC) fluorimetric reagent for primary amines
2,3-Naphthalenedialdehyde (NDA)
Formel:
C₁₂H₈O₂ MW: 184,19 g/mol |
MDL nummer:
MFCD00016618 CAS-nummer: 7149-49-7 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 131.0 to 135.0 deg-C |
Suitability for amin-analysis | to pass test |
Effect test of HPLC derivatization | Effective as labeling agent for Histamine |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...