5-(Benzoylamino)valeric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI

B0101-1G B0101-100MG B0101-5G
TCIAB0101-1GEA 1100 NOK
TCIAB0101-1G TCIAB0101-100MG TCIAB0101-5G
5-(Benzoylamino)valeric acid ≥98.0% (by GC, titration analysis)
5-(Benzoylamino)valeric acid
Formel: C₁₂H₁₅NO₃
MW: 221,26 g/mol
MDL nummer: MFCD00021790
CAS-nummer: 15647-47-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 92.0 to 95.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR