Nonyl chloroformate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Chloroformic Acid Nonyl Ester, Chloroformicacidn-nonylester

B0262-25ML B0262-5ML
TCIAB0262-25MLEA 1400 NOK
TCIAB0262-25ML TCIAB0262-5ML
Nonyl chloroformate ≥95.0% (by titrimetric analysis)
Nonyl chloroformate
Formel: C₁₀H₁₉ClO₂
MW: 206,71 g/mol
MDL nummer: MFCD00059485
CAS-nummer: 57045-82-6
FN: 3277
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR