meso-Butane-1,2,3,4-tetracarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: trans-Butane-1,2,3,4-tetracarboxylic acid

B0684-25G B0684-500G
TCIAB0684-25GEA 220 NOK
TCIAB0684-25G TCIAB0684-500G
meso-Butane-1,2,3,4-tetracarboxylic acid ≥98.0% (by GC, titration analysis)
meso-Butane-1,2,3,4-tetracarboxylic acid
Formel: C₈H₁₀O₈
MW: 234,16 g/mol
MDL nummer: MFCD00070496
CAS-nummer: 4534-68-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR