N-Butyl-p-toluenesulphonamide ≥97.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: N-Butyl-4-methylbenzenesulphonamide

B0745-25G B0745-500G
TCIAB0745-25GEA 365 NOK
TCIAB0745-25G TCIAB0745-500G
N-Butyl-p-toluenesulphonamide ≥97.0% (by total nitrogen basis)
N-Butyl-p-toluenesulphonamide
Formel: C₁₁H₁₇NO₂S
MW: 227,32 g/mol
Kokepunkt: 234°C
MDL nummer: MFCD00027162
CAS-nummer: 1907-65-9
EINECS: 217-612-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 38.0 to 42.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR