4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 4-(tert-butyl)cyclohexanecarboxylic acid
B1285-25G B1285-5G
TCIAB1285-25GEA 2850 NOK
TCIAB1285-25G TCIAB1285-5G
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₁H₂₀O₂
MW: 184,28 g/mol
MDL nummer: MFCD00042622
CAS-nummer: 5451-55-8
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %