Bis(2-dimethylaminoethyl)ether ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Dimethylaminoethyl Ether, 2,2'-Oxybis(N,N-dimethylethanamine)

B1291-25ML B1291-500ML
TCIAB1291-25MLEA 220 NOK
TCIAB1291-25ML TCIAB1291-500ML
Bis(2-dimethylaminoethyl)ether ≥98.0% (by GC)
Bis(2-dimethylaminoethyl)ether
Formel: C₈H₂₀N₂O
MW: 160,26 g/mol
Kokepunkt: 80°C
Flash punkt: 56°C
MDL nummer: MFCD00059199
CAS-nummer: 3033-62-3
EINECS: 221-220-5
FN: 2929
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR