1,4-Benzenedimethanethiol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene, 1,4-BDMT

B1521-1G B1521-5G
TCIAB1521-1GEA 660 NOK
TCIAB1521-1G TCIAB1521-5G
1,4-Benzenedimethanethiol ≥98.0% (by GC)
1,4-Benzenedimethanethiol
Formel: C₈H₁₀S₂
MW: 170,29 g/mol
Kokepunkt: 156°C
MDL nummer: MFCD00004872
CAS-nummer: 105-09-9
EINECS: 203-269-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR