2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI

B1535-1G B1535-100MG
TCIAB1535-1GEA 7275 NOK
TCIAB1535-1G TCIAB1535-100MG
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide ≥97.0% (by HPLC, titration analysis)
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide
Formel: C₁₈H₁₉NO₂S₂
MW: 345,48 g/mol
MDL nummer: MFCD00142790
CAS-nummer: 220191-36-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR