Ethyl-3-(benzylamino)propionate ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: N-Benzyl-beta-alanine Ethyl Ester, N-Benzyl-3-aminopropionic Acid Ethyl Ester, Ethyl 3-(benzylamino)propanoate

B1579-25ML B1579-500ML
TCIAB1579-25MLEA 620 NOK
TCIAB1579-25ML TCIAB1579-500ML
Ethyl-3-(benzylamino)propionate ≥95.0% (by GC)
Ethyl-3-(benzylamino)propionate
Formel: C₁₂H₁₇NO₂
MW: 207,27 g/mol
Kokepunkt: 135°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00085541
CAS-nummer: 23583-21-3
EINECS: 245-759-0

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR