(3R)-(-)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0% (by GC)
Leverandør: TCI
|
Danger
|
B1580-5G
B1580-1G
TCIAB1580-5GEA
5025
NOK
TCIAB1580-5G
TCIAB1580-1G
(3R)-(-)-1-Benzyl-3-(ethylamino)pyrrolidine ≥95.0% (by GC)
(3R)-(-)-1-Benzyl-3-(ethylamino)pyrrolidine
Formel:
C₁₃H₂₀N₂ MW: 204,32 g/mol |
MDL nummer:
MFCD00191306 CAS-nummer: 376591-05-8 FN: 2735 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Specific rotation [a]20/D | -6.0 to -9.0 deg (C=10, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...