N-(tert-Butoxycarbonyl)-p-toluenesulphonamide ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: N-Boc-p-toluenesulphonamide, tert-Butyl tosylcarbamate

B1648-10G B1648-25G
TCIAB1648-10GEA 1100 NOK
TCIAB1648-10G TCIAB1648-25G
N-(tert-Butoxycarbonyl)-p-toluenesulphonamide ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-p-toluenesulphonamide
Formel: C₁₂H₁₇NO₄S
MW: 271,33 g/mol
MDL nummer: MFCD00134267
CAS-nummer: 18303-04-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR