2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone

Leverandør: TCI

Synonymer: Dimethyl 3,4-Diphenylcyclopentadienone-2,5-dicarboxylate, 3,4-Diphenylcyclopentadienone-2,5-dicarboxylic Acid Dimethyl Ester

B1962-1G B1962-5G
TCIAB1962-1GEA 1500 NOK
TCIAB1962-1G TCIAB1962-5G
2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
Formel: C₂₁H₁₆O₅
MW: 348,35 g/mol
MDL nummer: MFCD00111095
CAS-nummer: 16691-79-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR