Diethyl (5-bromopentyl)malonate ≥97.0% (by GC)
Leverandør: TCI
Synonymer: (5-Bromopentyl)malonic Acid Diethyl Ester
B2083-25G B2083-5G
TCIAB2083-25GEA 3450 NOK
TCIAB2083-25G TCIAB2083-5G
Diethyl (5-bromopentyl)malonate ≥97.0% (by GC)
Diethyl (5-bromopentyl)malonate
Formel: C₁₂H₂₁BrO₄
MW: 309,20 g/mol
Kokepunkt: 161°C
MDL nummer: MFCD00048565
CAS-nummer: 1906-95-2
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %