N-(tert-Butoxycarbonyl)-2-nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
N-Boc-2-nitrobenzenesulphonamide
B2303-1G
B2303-25G
B2303-5G
TCIAB2303-1GEA
485
NOK
TCIAB2303-1G
TCIAB2303-25G
TCIAB2303-5G
N-(tert-Butoxycarbonyl)-2-nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-2-nitrobenzenesulphonamide
Formel:
C₁₁H₁₄N₂O₆S MW: 302,30 g/mol |
MDL nummer:
MFCD06796217 CAS-nummer: 198572-71-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...