1-Boc-isonipecotic acid ethyl ester ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate
B2740-5G
B2740-25G
TCIAB2740-5GEA
285
NOK
TCIAB2740-5G
TCIAB2740-25G
1-Boc-isonipecotic acid ethyl ester ≥97.0% (by GC)
1-Boc-isonipecotic acid ethyl ester
Formel:
C₁₃H₂₃NO₄ MW: 257,33 g/mol Kokepunkt: 140°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD01763998 CAS-nummer: 142851-03-4 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid to slightly cloudy liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...