5-Bromo-2,3-pyridinediol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
5-Bromopyridine-2,3-diol, 5-Bromo-2,3-dihydroxypyridine
B2780-1G
B2780-25G
B2780-5G
TCIAB2780-1GEA
540
NOK
TCIAB2780-1G
TCIAB2780-25G
TCIAB2780-5G
5-Bromo-2,3-pyridinediol ≥98.0% (by GC, titration analysis)
5-Bromo-2,3-pyridinediol
Formel:
C₅H₄BrNO₂ MW: 190,00 g/mol Smeltepunkt: 249°C |
MDL nummer:
MFCD08276314 CAS-nummer: 34206-49-0 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...