Bis(4-tert-butylphenyl)amine ≥90.0% (by GC)
Leverandør: TCI
Synonymer:
4,4'-Di-tert-butyldiphenylamine
B2803-25G
B2803-500G
TCIAB2803-25GEA
220
NOK
TCIAB2803-25G
TCIAB2803-500G
Bis(4-tert-butylphenyl)amine ≥90.0% (by GC)
Bis(4-tert-butylphenyl)amine
Formel:
C₂₀H₂₇N MW: 281,44 g/mol Kokepunkt: 195°C |
MDL nummer:
MFCD00417391 CAS-nummer: 4627-22-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 90.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...