4-Butylbenzyl bromide ≥97.0% (by GC)

Leverandør: TCI
Danger

B3005-1G B3005-5G
TCIAB3005-1GEA 890 NOK
TCIAB3005-1G TCIAB3005-5G
4-Butylbenzyl bromide ≥97.0% (by GC)
4-Butylbenzyl bromide
Formel: C₁₁H₁₅Br
MW: 227,15 g/mol
Kokepunkt: 144°C
CAS-nummer: 10531-16-5
FN: 3265
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR