N-Benzyl-N-nitroso-p-toluenesulphonamide ≥98.0% (by HPLC)

Leverandør: TCI
Warning

B3051-25G B3051-5G
TCIAB3051-25GEA 5975 NOK
TCIAB3051-25G TCIAB3051-5G
N-Benzyl-N-nitroso-p-toluenesulphonamide ≥98.0% (by HPLC)
N-Benzyl-N-nitroso-p-toluenesulphonamide
Formel: C₁₄H₁₄N₂O₃S
Smeltepunkt: 85°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00216617
CAS-nummer: 33528-13-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR