O-tert-Butyl-L-tyrosine ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: H-Tyr(tBu)-OH

B3212-1G B3212-5G
TCIAB3212-1GEA 230 NOK
TCIAB3212-1G TCIAB3212-5G
O-tert-Butyl-L-tyrosine ≥98.0% (by HPLC)
O-tert-Butyl-L-tyrosine
Formel: C₁₃H₁₉NO₃
MW: 237,30 g/mol
MDL nummer: MFCD00037202
CAS-nummer: 18822-59-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D -9.0 to -4.0 deg (C=1, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR