tert-Butyl propionate ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Propionic Acid tert-Butyl Ester

B3327-100G B3327-25G
TCIAB3327-100GEA 3000 NOK
TCIAB3327-100G TCIAB3327-25G
tert-Butyl propionate ≥98.0% (by GC)
tert-Butyl propionate
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokepunkt: 119°C
Flash punkt: 21°C
MDL nummer: MFCD00009304
CAS-nummer: 20487-40-5
FN: 1914
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR