2-Bromodibenzothiophene ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 2-Bromodibenzo[b,d]thiophene

B3525-25G B3525-5G
TCIAB3525-25GEA 1575 NOK
TCIAB3525-25G TCIAB3525-5G
2-Bromodibenzothiophene ≥95.0% (by GC)
2-Bromodibenzothiophene
Formel: C₁₂H₇BrS
MW: 263,15 g/mol
MDL nummer: MFCD00089285
CAS-nummer: 22439-61-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 123.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR