2-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2-Bromo-9,9'-spirobi[fluorene]

B3560-5G B3560-1G
TCIAB3560-5GEA 485 NOK
TCIAB3560-5G TCIAB3560-1G
2-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
2-Bromo-9,9'-spirobi[9H-fluorene]
Formel: C₂₅H₁₅Br
MW: 395,30 g/mol
MDL nummer: MFCD08704217
CAS-nummer: 171408-76-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR