1,3,5-Benzenetrimethanol ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 1,3,5-Tris(hydroxymethyl)benzene, Benzene-1,3,5-triyltrimethanol

B3586-1G B3586-5G
TCIAB3586-1GEA 1150 NOK
TCIAB3586-1G TCIAB3586-5G
1,3,5-Benzenetrimethanol ≥95.0% (by GC)
1,3,5-Benzenetrimethanol
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
MDL nummer: MFCD01108484
CAS-nummer: 4464-18-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR