5-Bromo-2-chloro-3-methylpyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
5-Bromo-2-chloro-3-picoline
B3744-5G
B3744-25G
TCIAB3744-5GEA
620
NOK
TCIAB3744-5G
TCIAB3744-25G
5-Bromo-2-chloro-3-methylpyridine ≥98.0% (by GC)
5-Bromo-2-chloro-3-methylpyridine
Formel:
C₆H₅BrClN MW: 206,47 g/mol Kokepunkt: 78°C |
MDL nummer:
MFCD03095093 CAS-nummer: 29241-60-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...