tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-BOC protected
B3787-25G
B3787-5G
TCIAB3787-25GEA
1775
NOK
TCIAB3787-25G
TCIAB3787-5G
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate
Formel:
C₁₁H₂₁NO₃ MW: 215,29 g/mol |
MDL nummer:
MFCD03094733 CAS-nummer: 116574-71-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...