4-Bromopiperidine hydrobromide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

B3830-5G B3830-1G
TCIAB3830-5GEA 1100 NOK
TCIAB3830-5G TCIAB3830-1G
4-Bromopiperidine hydrobromide ≥98.0% (by titrimetric analysis)
4-Bromopiperidine hydrobromide
Formel: C₅H₁₁Br₂N
MW: 244.9600 g/mol
MDL nummer: MFCD00191858
CAS-nummer: 54288-70-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 190.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR