Boc-cycloleucine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
1-(Boc-amino)cyclopentanecarboxylic acid, 1-N-BOC-Aminocyclopentanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
B3831-1G
B3831-5G
TCIAB3831-1GEA
435
NOK
TCIAB3831-1G
TCIAB3831-5G
Boc-cycloleucine ≥98.0% (by GC, titration analysis)
Boc-cycloleucine
Formel:
C₁₁H₁₉NO₄ MW: 229,28 g/mol |
MDL nummer:
MFCD01076126 CAS-nummer: 35264-09-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...