2-Bromo-5-hydroxypyrazine ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Bromopyrazin-2-ol

B3881-200MG B3881-1G
TCIAB3881-200MGEA 1525 NOK
TCIAB3881-200MG TCIAB3881-1G
2-Bromo-5-hydroxypyrazine ≥95.0% (by GC)
2-Bromo-5-hydroxypyrazine
Formel: C₄H₃BrN₂O
MW: 174,9800 g/mol
MDL nummer: MFCD06245330
CAS-nummer: 374063-92-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR