Bis(4-bromophenyl)acetylene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1,2-Bis(4-bromophenyl)ethyne

B3996-1G B3996-5G
TCIAB3996-1GEA 650 NOK
TCIAB3996-1G TCIAB3996-5G
Bis(4-bromophenyl)acetylene ≥98.0% (by GC)
Bis(4-bromophenyl)acetylene
Formel: C₁₄H₈Br₂
MW: 336,03 g/mol
MDL nummer: MFCD00185147
CAS-nummer: 2789-89-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR