4-Bromo-1-methyl-1H-pyrazole ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
B4060-5G
B4060-25G
TCIAB4060-5GEA
245
NOK
TCIAB4060-5G
TCIAB4060-25G
4-Bromo-1-methyl-1H-pyrazole ≥98.0% (by GC)
4-Bromo-1-methyl-1H-pyrazole
Formel:
C₄H₅BrN₂ MW: 161,00 g/mol Kokepunkt: 78°C Flash punkt: 93°C |
MDL nummer:
MFCD02179565 CAS-nummer: 15803-02-8 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...