N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane ≥90.0% (by HPLC)
Leverandør: TCI
Synonymer:
BCN-POE3-NH2, BCN-amine, rel-((1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-yl)methyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate, endo-BCN-PEG2-NH2
B4062-25MG
B4062-100MG
TCIAB4062-25MGEA
2350
NOK
TCIAB4062-25MG
TCIAB4062-100MG
N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane ≥90.0% (by HPLC)
N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane
Formel:
C₁₇H₂₈N₂O₄ MW: 324,42 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD19705418 CAS-nummer: 1263166-93-3 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) (CAD) | min. 90.0 area% |
Elemental analysis (Nitrogen) | 7.70 to 8.90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...