Benzo[b]thiophene-3-carboxaldehyde ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Benzo[b]thiophene-3-carbaldehyde
B4086-1G
B4086-5G
TCIAB4086-1GEA
340
NOK
TCIAB4086-1G
TCIAB4086-5G
Benzo[b]thiophene-3-carboxaldehyde ≥98.0% (by GC)
Benzo[b]thiophene-3-carboxaldehyde
Formel:
C₉H₆OS MW: 162,2083 g/mol Kokepunkt: 133°C |
MDL nummer:
MFCD00052376 CAS-nummer: 5381-20-4 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...