2-(Benzylthio)ethanol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2-Hydroxyethyl Benzyl Sulphide, 2-(Benzylthio)ethan-1-ol

B4087-5G B4087-25G
TCIAB4087-5GEA 220 NOK
TCIAB4087-5G TCIAB4087-25G
2-(Benzylthio)ethanol ≥98.0% (by GC)
2-(Benzylthio)ethanol
Formel: C₉H₁₂OS
MW: 168,25 g/mol
Kokepunkt: 120°C
MDL nummer: MFCD00014039
CAS-nummer: 3878-41-9
EINECS: 223-405-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR