Benzbromarone ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (3,5-Dibromo-4-hydroxyphenyl)(2-ethylbenzofuran-3-yl)methanone, 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran

B4099-1G B4099-5G
TCIAB4099-1GEA 485 NOK
TCIAB4099-1G TCIAB4099-5G
Benzbromarone ≥98.0% (by GC, titration analysis)
Benzbromarone
Formel: C₁₇H₁₂Br₂O₃
MW: 424,09 g/mol
MDL nummer: MFCD00078962
CAS-nummer: 3562-84-3
EINECS: 222-630-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,01065

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR