Benzyl-3-oxopiperazine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: Benzyl 3-oxopiperazine-1-carboxylate, 3-Oxopiperazine-1-carboxylic Acid Benzyl Ester, 1-Carbobenzoxy-3-oxopiperazine, 1-Cbz-3-oxopiperazine

B4100-1G B4100-5G
TCIAB4100-1GEA 475 NOK
TCIAB4100-1G TCIAB4100-5G
Benzyl-3-oxopiperazine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)
Benzyl-3-oxopiperazine-1-carboxylate
Formel: C₁₂H₁₄N₂O₃
MW: 234,26 g/mol
MDL nummer: MFCD00173924
CAS-nummer: 78818-15-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR