(S)-1,2-Butanediol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (S)-1,2-Butylene Glycol, (S)-1,2-Dihydroxybutane, (S)-Butane-1,2-diol

B4119-1G B4119-5G
TCIAB4119-1GEA 300 NOK
TCIAB4119-1G TCIAB4119-5G
(S)-1,2-Butanediol ≥98.0% (by GC)
(S)-1,2-Butanediol
Formel: C₄H₁₀O₂
MW: 90,12 g/mol
Kokepunkt: 55°C
MDL nummer: MFCD09953764
CAS-nummer: 73522-17-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -15.0 to -18.0 deg (C=3、EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR