N-Boc-4-oxo-L-proline ≥98.0% (by titrimetric analysis)

Leverandør: TCI

B4141-1G B4141-5G
TCIAB4141-1GEA 620 NOK
TCIAB4141-1G TCIAB4141-5G
N-Boc-4-oxo-L-proline ≥98.0% (by titrimetric analysis)
N-Boc-4-oxo-L-proline
Formel: C₁₀H₁₅NO₅
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01860669
CAS-nummer: 84348-37-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +19.0 to +22.0 deg (C=0.5,Acetone)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR