6-Bromo-m-toluic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
B4145-5G
B4145-25G
TCIAB4145-5GEA
380
NOK
TCIAB4145-5G
TCIAB4145-25G
6-Bromo-m-toluic acid ≥98.0% (by GC, titration analysis)
6-Bromo-m-toluic acid
Formel:
C₈H₇BrO₂ MW: 215,05 g/mol |
MDL nummer:
MFCD00079722 CAS-nummer: 6967-82-4 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 136.0 to 140.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...