2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
Benzo[1,2-b:4,5-b']dithiophene-2,6-diboronic Acid Bis(pinacol) Ester
B4158-1G
B4158-5G
TCIAB4158-1GEA
1925
NOK
TCIAB4158-1G
TCIAB4158-5G
2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by GC)
2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene
Formel:
C₂₂H₂₈B₂O₄S₂ MW: 442,20 g/mol |
CAS-nummer:
861398-06-3 |
|
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
![](/m-no.vwr.com/no_NO/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...