2-Bromo-3-nitropyridine ≥98.0% (by GC)

Leverandør: TCI
Warning

B4193-1G B4193-5G
TCIAB4193-1GEA 325 NOK
TCIAB4193-1G TCIAB4193-5G
2-Bromo-3-nitropyridine ≥98.0% (by GC)
2-Bromo-3-nitropyridine
Formel: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL nummer: MFCD00955613
CAS-nummer: 19755-53-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR