N-Boc-m-phenylenediamine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic acid tert-Butyl ester, N-Boc-1,3-phenylenediamine, 3-(tert-Butoxycarbonylamino)aniline, N-(tert-Butoxycarbonyl)-1,3-phenylenediamine, tert-Butyl (3-aminophenyl)carbamate, 3-(Boc-amino)aniline
B4222-1G
B4222-5G
TCIAB4222-1GEA
380
NOK
TCIAB4222-1G
TCIAB4222-5G
N-Boc-m-phenylenediamine ≥98.0% (by GC, titration analysis)
N-Boc-m-phenylenediamine
Formel:
C₁₁H₁₆N₂O₂ MW: 208,261 g/mol |
MDL nummer:
MFCD06657915 CAS-nummer: 68621-88-5 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...