4-Bromo-4'-chlorobenzophenone ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (4-Bromophenyl)(4-chlorophenyl)methanone

B4224-1G B4224-5G
TCIAB4224-1GEA 1675 NOK
TCIAB4224-1G TCIAB4224-5G
4-Bromo-4'-chlorobenzophenone ≥98.0% (by GC)
4-Bromo-4'-chlorobenzophenone
Formel: C₁₃H₈BrClO
MW: 295,5590 g/mol
MDL nummer: MFCD00045208
CAS-nummer: 27428-57-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18e7a92499d622cbb7fa5da0a3d1b986462d2008&vid=18e7a92499d622cbb7fa5da0a3d1b986462d2008&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR